Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007192.3 | NP_009123.1 | 9 | 1422 | |
XM_011536381.2 | XP_011534683.1 | 10 | 1424 | ||
GenBank mRNA | AK001719.1 | - | 9 | 1158 | |
AK024072.1 | - | 9 | 1424 | ||
BC000565.2 | - | 9 | 1159 | ||
BC014046.2 | - | 9 | 1159 | ||
BC064561.1 | - | 9 | 1147 | ||
BC073849.1 | - | 9 | 1156 | ||
BC111402.1 | - | 9 | 1171 |