Gene Transcripts

Gene Symbol
ACAD9
Entrez Gene ID
28976
Gene Name
acyl-CoA dehydrogenase family member 9
Gene Aliases
NPD002
Chromosome Location
Chr. 3: 128879490 - 128916075 on Build GRCh38
UniGene ID
Hs.567482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014049.4 NP_054768.2 8 1074
NR_033426.1 8 1250
XM_017006264.1 XP_016861753.1 8 1238
XM_017006265.1 XP_016861754.1 2 220
GenBank mRNA AB209427.1 - 8 2767
AF327351.1 - 8 904
AK022568.1 - 7 1000
AK024012.1 - 8 918
AK130311.1 - 8 945
AK294597.1 - 8 742
AK297952.1 - 8 1118
AK310644.1 - 7 847
BC001817.2 - 1 38
BC007970.2 - 8 951 4304209
BC013354.2 - 8 928 3935925

View Full Product Details