Gene Transcripts
- Gene Symbol
- TEX14
- Entrez Gene ID
- 56155
- Gene Name
- testis expressed 14, intercellular bridge forming factor
- Gene Aliases
- CT113
- Chromosome Location
- Chr. 17: 58556677 - 58692055 on Build GRCh38
- UniGene ID
- Hs.390221
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001201457.1 |
NP_001188386.1
|
13, 14 |
1815 |
|
NM_031272.4 |
NP_112562.3
|
13, 14 |
1797 |
|
NM_198393.3 |
NP_938207.2
|
13, 14 |
1797 |
|
XM_011525029.2 |
XP_011523331.1
|
13, 14 |
1776 |
|
XM_017024860.1 |
XP_016880349.1
|
11, 12 |
1447 |
|
XM_017024861.1 |
XP_016880350.1
|
12, 13 |
1622 |
|
GenBank mRNA |
AF285601.1 |
- |
2, 3 |
261 |
|
AK128217.1 |
- |
9, 10 |
1232 |
|
AK308299.1 |
- |
13, 14 |
1757 |
|
AL834143.1 |
- |
13, 14 |
1796 |
|
BC040526.1 |
- |
13, 14 |
1783 |
4831308 |
View Full Product Details