Gene Transcripts

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
Chromosome Location
Chr. 6: 12256464 - 12297194 on Build GRCh38
UniGene ID
Hs.511899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168319.1 NP_001161791.1 3, 4 715
NM_001955.4 NP_001946.3 3, 4 718
XM_011514330.2 XP_011512632.1 4, 5 474
XM_011514331.2 XP_011512633.1 5, 6 635
XM_011514332.2 XP_011512634.1 4, 5 472
XM_017010331.1 XP_016865820.1 4, 5 658
GenBank mRNA AK226096.1 - 3, 4 633
AK291838.1 - 3, 4 655
AK313914.1 - 3, 4 650
BC009720.1 - 3, 4 587 3900704
BX439515.2 - 3, 4 678
CR541903.1 - 3, 4 384
S56805.1 - 3, 4 720
Y00749.1 - 3, 4 636

View Full Product Details