Gene Transcripts

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
Chromosome Location
Chr. 1: 7784285 - 7845181 on Build GRCh38
UniGene ID
Hs.162200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289861.1 NP_001276790.1 17 2431
NM_001289862.1 NP_001276791.1 17 2431
NM_001289863.1 NP_001276792.1 17 2410
NM_001289864.1 NP_001276793.1 17 2492
NM_016831.2 NP_058515.1 17 2407
XM_005263524.3 XP_005263581.1 16 2306
XM_011542385.2 XP_011540687.1 16 2306
XM_011542390.2 XP_011540692.1 16 2285
XM_017002723.1 XP_016858212.1 16 2303
XM_017002724.1 XP_016858213.1 16 2303
XM_017002725.1 XP_016858214.1 16 2285
XM_017002726.1 XP_016858215.1 16 2282
XM_017002727.1 XP_016858216.1 18 2625
XM_017002728.1 XP_016858217.1 16 2367
XM_017002729.1 XP_016858218.1 14 1807
XM_017002730.1 XP_016858219.1 16 2367
XM_017002731.1 XP_016858220.1 16 2346
XM_017002732.1 XP_016858221.1 15 2395
XM_017002733.1 XP_016858222.1 18 2532
XM_017002734.1 XP_016858223.1 19 2674
XM_017002735.1 XP_016858224.1 18 2532
XM_017002736.1 XP_016858225.1 19 2653
XM_017002737.1 XP_016858226.1 13 1790
XM_017002738.1 XP_016858227.1 13 1796
GenBank mRNA AB047686.1 - 16 2391
AK307760.1 - 16 2285
AK308303.1 - 17 2480
AL157954.1 - 16 2318
AY493418.1 - 16 2214
BC144137.1 - 16 2396
BC146781.1 - 16 2391 9007235

View Full Product Details