Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017429.2 | NP_059125.2 | 10, 11 | 1633 | |
XM_011523109.1 | XP_011521411.1 | 8, 9 | 1481 | ||
XM_017023289.1 | XP_016878778.1 | 6, 7 | 747 | ||
GenBank mRNA | AF294900.1 | - | 10, 11 | 1633 | |
AK001592.1 | - | 10, 11 | 1633 | ||
AK296011.1 | - | 9, 10 | 1505 | ||
BC126210.1 | - | 10, 11 | 1535 | 8991926 | |
BC126212.1 | - | 10, 11 | 1535 | 8991928 |