Gene Transcripts

Gene Symbol
TRIM56
Entrez Gene ID
81844
Gene Name
tripartite motif containing 56
Gene Aliases
RNF109
Chromosome Location
Chr. 7: 101085424 - 101097967 on Build GRCh38
UniGene ID
Hs.521092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030961.2 NP_112223.1 3 2372
XM_011516589.2 XP_011514891.1 2 2224
GenBank mRNA AK075255.1 - 1 792
AL512757.1 - 1 362
BC005847.2 - 3 2372 2907142
BC048194.1 - 3 2306 5814702

View Full Product Details