Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000373.3 | NP_000364.1 | 3 | 537 | |
NR_033434.1 | 2 | 383 | |||
NR_033437.1 | 4 | 636 | |||
GenBank mRNA | AB062285.1 | - | 3 | 459 | |
AK291306.1 | - | 3 | 453 | ||
AL080099.2 | - | 2 | 316 | ||
BC000364.2 | - | 3 | 454 | 2964556 | |
BC007511.2 | - | 3 | 454 | 2964556 | |
CR456787.1 | - | 3 | 431 | ||
D86227.1 | - | 3 | 450 | ||
D86228.1 | - | 3 | 450 | ||
D86230.1 | - | 3 | 450 | ||
EU921886.1 | - | 3 | 451 | ||
EU921887.1 | - | 2 | 297 | ||
EU921888.1 | - | 2 | 297 | ||
EU921889.1 | - | 3 | 396 | ||
EU921890.1 | - | 3 | 451 | ||
EU921891.1 | - | 4 | 550 | ||
EU921892.1 | - | 3 | 451 | ||
EU921894.1 | - | 3 | 451 | ||
EU921895.1 | - | 2 | 297 | ||
J03626.1 | - | 3 | 535 | ||
KU178375.1 | - | 3 | 431 | ||
KU178376.1 | - | 4 | 530 | ||
M36661.1 | - | 3 | 397 |