Gene Transcripts

Gene Symbol
HSPA6
Entrez Gene ID
3310
Gene Name
heat shock protein family A (Hsp70) member 6
Gene Aliases
HSP70B'
Chromosome Location
Chr. 1: 161524540 - 161526897 on Build GRCh38
UniGene ID
Hs.654614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002155.4 NP_002146.2 1 1611
GenBank mRNA AK093925.1 - 1 1611
AK223362.1 - 1 1611
AK295212.1 - 2 1534
AK312756.1 - 1 1611
BC035665.1 - 1 1601 5723718
X51758.1 - 1 430

Gene Symbol
HSPA7
Entrez Gene ID
3311
Gene Name
heat shock protein family A (Hsp70) member 7
Gene Aliases
HSP70B
Chromosome Location
Chr. 1: 161606059 - 161608551 on Build GRCh38
UniGene ID
Hs.662180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024151.1 1 1725

View Full Product Details