Gene Transcripts

Gene Symbol
CLK4
Entrez Gene ID
57396
Gene Name
CDC like kinase 4
Gene Aliases
Chromosome Location
Chr. 5: 178602664 - 178627053 on Build GRCh38
UniGene ID
Hs.406557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020666.2 NP_065717.1 2 238
GenBank mRNA AF294429.1 - 1 130
AK294921.1 - 2 237
AK304614.1 - 2 240
AK308578.1 - 2 271
AK308674.1 - 2 297
AK308879.1 - 2 267
AK309215.1 - 2 267
AK310427.1 - 2 220
AK311485.1 - 2 240
BC015942.1 - 2 238
BC034633.1 - 2 192
BC065732.1 - 2 219
BC136261.1 - 2 186 9020248
BC143547.1 - 2 186 9052058
BC151233.1 - 2 273
CR749504.1 - 2 214

View Full Product Details