Gene Transcripts
- Gene Symbol
- KPNA3
- Entrez Gene ID
- 3839
- Gene Name
- karyopherin subunit alpha 3
- Gene Aliases
- IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
- Chromosome Location
- Chr. 13: 49699307 - 49792921 on Build GRCh38
- UniGene ID
- Hs.527919
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002267.3 |
NP_002258.2
|
5, 6 |
702 |
|
XM_017020561.1 |
XP_016876050.1
|
5, 6 |
385 |
|
GenBank mRNA |
AF005263.1 |
- |
5, 6 |
299 |
|
AF034756.1 |
- |
5, 6 |
289 |
|
AK223480.1 |
- |
5, 6 |
324 |
|
AK290528.1 |
- |
5, 6 |
362 |
|
AK291000.1 |
- |
5, 6 |
326 |
|
BC017355.2 |
- |
5, 6 |
311 |
5020312 |
BC024202.2 |
- |
5, 6 |
311 |
3531069 |
BC035090.2 |
- |
5, 6 |
703 |
5261465 |
D89618.1 |
- |
5, 6 |
378 |
|
DN991804.1 |
- |
5, 6 |
490 |
|
Y12394.1 |
- |
5, 6 |
302 |
|
View Full Product Details