Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003818.3 | NP_003809.1 | 7 | 939 | |
XM_006723660.1 | XP_006723723.1 | 7 | 930 | ||
XM_011529390.1 | XP_011527692.1 | 8 | 1262 | ||
GenBank mRNA | AF069532.1 | - | 6 | 535 | |
AK001414.1 | - | 7 | 660 | ||
AK027695.1 | - | 7 | 646 | ||
AK091045.1 | - | 8 | 1262 | ||
AK095430.1 | - | 4 | 863 | ||
AK303426.1 | - | 3 | 213 | ||
AK315489.1 | - | 7 | 920 | ||
BC025751.1 | - | 7 | 660 | 5199748 | |
Y16521.1 | - | 7 | 865 |