Gene Transcripts

Gene Symbol
CHST4
Entrez Gene ID
10164
Gene Name
carbohydrate sulfotransferase 4
Gene Aliases
GST3, GlcNAc6ST2, HECGLCNAC6ST, LSST
Chromosome Location
Chr. 16: 71526120 - 71538590 on Build GRCh38
UniGene ID
Hs.251383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166395.1 NP_001159867.1 2 917
NM_005769.2 NP_005760.1 2 865
GenBank mRNA AF131235.1 - 2 835
AF149783.1 - 1 746
AF280088.1 - 2 775
AK026635.1 - 2 725
AK074746.1 - 2 722
BC035282.1 - 2 731 5179826

View Full Product Details