Gene Transcripts

Gene Symbol
CHST6
Entrez Gene ID
4166
Gene Name
carbohydrate sulfotransferase 6
Gene Aliases
MCDC1
Chromosome Location
Chr. 16: 75473124 - 75495419 on Build GRCh38
UniGene ID
Hs.655622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021615.4 NP_067628.1 3 6864
GenBank mRNA AF280086.1 - 4 3740
BC036640.1 - 1 1133
BC041645.1 - 1 1133

View Full Product Details