Gene Transcripts

Gene Symbol
RTN4RL1
Entrez Gene ID
146760
Gene Name
reticulon 4 receptor like 1
Gene Aliases
NGRH2, NgR3
Chromosome Location
Chr. 17: 1934677 - 2025345 on Build GRCh38
UniGene ID
Hs.22917
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178568.3 NP_848663.1 2 1421
GenBank mRNA AF532859.1 - 1 941
AL834409.1 - 1 887
AY250219.1 - 1 941
BC128608.1 - 2 963 40121824

View Full Product Details