Gene Transcripts

Gene Symbol
ITGB3
Entrez Gene ID
3690
Gene Name
integrin subunit beta 3
Gene Aliases
BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT
Chromosome Location
Chr. 17: 47253842 - 47312711 on Build GRCh38
UniGene ID
Hs.218040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000212.2 NP_000203.2 15 3053
GenBank mRNA J02703.1 - 15 3053
M20311.1 - 15 3049
M35999.1 - 15 3051

View Full Product Details