Gene Transcripts

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
Chromosome Location
Chr. 7: 28995630 - 29146537 on Build GRCh38
UniGene ID
Hs.233389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019029.2 NP_061902.2 9 1022
NM_031311.3 NP_112601.3 9 934
XM_011515437.1 XP_011513739.1 9 1101
XM_017012366.1 XP_016867855.1 9 934
GenBank mRNA AF106704.2 - 9 854
AF217508.1 - 8 821
AF282617.1 - 9 884
AK075433.1 - 9 935
AK124472.1 - 13 1462
AK225545.1 - 9 885
AY358549.1 - 9 911
BC016838.2 - 9 982 3888647

View Full Product Details