Gene Transcripts

Gene Symbol
MFRP
Entrez Gene ID
83552
Gene Name
membrane frizzled-related protein
Gene Aliases
MCOP5, NNO2, RD6
Chromosome Location
Chr. 11: 119338934 - 119346673 on Build GRCh38
UniGene ID
Hs.632102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031433.3 NP_113621.1 9 1224
GenBank mRNA AB055505.1 - 9 1211
AJ862823.1 - 9 1206
AK055132.1 - 9 1224

Gene Symbol
C1QTNF5
Entrez Gene ID
114902
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
CTRP5
Chromosome Location
Chr. 11: 119338934 - 119346673 on Build GRCh38
UniGene ID
Hs.632102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015645.4 NP_056460.1 9 1224

View Full Product Details