Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr. 17: 44759037 - 44781846 on Build GRCh38
UniGene ID
Hs.6088
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002390.5 NP_002381.2 8 688
XM_005257373.3 XP_005257430.1 8 830
GenBank mRNA AB009675.1 - 8 667
D17390.1 - 5 397

View Full Product Details