Gene Transcripts

Gene Symbol
HTRA4
Entrez Gene ID
203100
Gene Name
HtrA serine peptidase 4
Gene Aliases
Chromosome Location
Chr. 8: 38974149 - 38988662 on Build GRCh38
UniGene ID
Hs.661014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153692.3 NP_710159.1 9 1438
GenBank mRNA AK075205.1 - 9 1409
AK300197.1 - 9 1317
BC057765.1 - 9 1370 30348811

View Full Product Details