Gene Transcripts

Gene Symbol
USP12
Entrez Gene ID
219333
Gene Name
ubiquitin specific peptidase 12
Gene Aliases
UBH1, USP12L1
Chromosome Location
Chr. 13: 27066150 - 27171896 on Build GRCh38
UniGene ID
Hs.42400
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182488.3 NP_872294.2 6, 7 985
XM_005266282.2 XP_005266339.1 6, 7 899
XM_011534956.2 XP_011533258.1 6, 7 736
XM_017020416.1 XP_016875905.1 5, 6 656
GenBank mRNA AF022789.1 - 6, 7 835
AK289685.1 - 6, 7 899
BC026072.1 - 6, 7 986 4830251

View Full Product Details