Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002307.3 | NP_002298.1 | 3 | 210 | |
GenBank mRNA | BP225960.1 | - | 3 | 210 | |
CR456917.1 | - | 2 | 185 | ||
CV575179.1 | - | 3 | 198 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042507.3 | NP_001035972.1 | 3 | 202 | |
GenBank mRNA | BC042911.2 | - | 2 | 178 | |
BC061588.1 | - | 3 | 201 | 6202380 | |
BC073743.1 | - | 3 | 201 | 5478866 | |
L07769.1 | - | 3 | 203 | ||
U06643.1 | - | 3 | 214 |