Gene Transcripts

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
Chromosome Location
Chr. 4: 41744082 - 41748970 on Build GRCh38
UniGene ID
Hs.87202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003924.3 NP_003915.2 1 503
GenBank mRNA BC017199.2 - 1 239
D82344.1 - 1 503

View Full Product Details