Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001455.3 | NP_001446.1 | 2 | 1926 | |
NM_201559.2 | NP_963853.1 | 3 | 1899 | ||
XM_005266867.4 | XP_005266924.1 | 3 | 1147 | ||
XM_005266868.3 | XP_005266925.1 | 2 | 1171 | ||
XM_011535626.1 | XP_011533928.1 | 2 | 1097 | ||
XM_011535628.2 | XP_011533930.1 | 2 | 5455 | ||
XM_011535629.2 | XP_011533931.1 | 2 | 1120 | ||
XM_017010585.1 | XP_016866074.1 | 3 | 2832 | ||
XM_017010586.1 | XP_016866075.1 | 2 | 2715 | ||
GenBank mRNA | AF032886.1 | - | 2 | 2507 | |
AJ001589.1 | - | 2 | 1643 | ||
AK092357.1 | - | 3 | 1287 | ||
AK122861.1 | - | 3 | 1482 | ||
AK301304.1 | - | 2 | 1088 | ||
AK303933.1 | - | 2 | 1072 | ||
AK311587.1 | - | 2 | 1186 | ||
BC020227.1 | - | 3 | 1887 | 4111403 | |
BC021224.2 | - | 3 | 1887 | 4137370 | |
BC068552.1 | - | 2 | 1926 | 30347471 | |
CR749261.1 | - | 2 | 1156 |