Gene Transcripts

Gene Symbol
ID2
Entrez Gene ID
3398
Gene Name
inhibitor of DNA binding 2, HLH protein
Gene Aliases
GIG8, ID2A, ID2H, bHLHb26
Chromosome Location
Chr. 2: 8681983 - 8684453 on Build GRCh38
UniGene ID
Hs.180919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002166.4 NP_002157.2 3 655
GenBank mRNA AK222682.1 - 3 582
BC030639.2 - 3 582 4820416
D13891.1 - 3 568
M97796.1 - 3 580

View Full Product Details