Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100816.1 | NP_001094286.1 | 2 | 696 | |
NM_004270.4 | NP_004261.1 | 2 | 412 | ||
GenBank mRNA | AF031383.1 | - | 2 | 370 | |
AF104251.1 | - | 1 | 304 | ||
AK308393.1 | - | 2 | 377 | ||
AV711090.1 | - | 2 | 424 | ||
BC005250.1 | - | 2 | 366 | ||
BC006968.1 | - | 2 | 366 | ||
BT007368.1 | - | 1 | 304 | ||
CR457009.1 | - | 1 | 304 | ||
CR541696.1 | - | 1 | 304 |