Gene Transcripts

Gene Symbol
SLC7A9
Entrez Gene ID
11136
Gene Name
solute carrier family 7 member 9
Gene Aliases
BAT1, CSNU3
Chromosome Location
Chr. 19: 32830511 - 32870958 on Build GRCh38
UniGene ID
Hs.743345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126335.1 NP_001119807.1 11 1213
NM_001243036.1 NP_001229965.1 11 1308
NM_014270.4 NP_055085.1 11 1299
XM_006722992.1 XP_006723055.1 9 974
XM_011526402.2 XP_011524704.1 11 2403
XM_017026230.1 XP_016881719.1 9 949
GenBank mRNA AB033548.1 - 11 1184
AF141289.1 - 11 1281
AJ249199.1 - 10 1158
AK026446.1 - 11 1282
AK223147.1 - 11 1287
AK313708.1 - 11 1200
AL365340.1 - 4 351
AL365341.1 - 3 135
AY170373.1 - 6 630
BC017962.1 - 11 1288 4608486

View Full Product Details