Gene Transcripts

Gene Symbol
SCML4
Entrez Gene ID
256380
Gene Name
sex comb on midleg-like 4 (Drosophila)
Gene Aliases
dJ47M23.1
Chromosome Location
Chr. 6: 107697297 - 107845959 on Build GRCh38
UniGene ID
Hs.486109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011535707.2 XP_011534009.1 6 1740
GenBank mRNA AK074125.1 - 4 3046
AK092812.1 - 2 918
BC033286.1 - 6 1488 4509724

View Full Product Details