Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003747.2 | NP_003738.2 | 1 | 291 | |
XM_006716263.3 | XP_006716326.1 | 1 | 997 | ||
XM_011543845.2 | XP_011542147.1 | 1 | 997 | ||
XM_011543846.2 | XP_011542148.1 | 1 | 997 | ||
XM_011543847.2 | XP_011542149.1 | 1 | 996 | ||
GenBank mRNA | AF082556.1 | - | 1 | 291 | |
AK301798.1 | - | 1 | 212 | ||
AK302031.1 | - | 1 | 312 | ||
BC098394.1 | - | 1 | 314 | 4374347 |