Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012089.2 | NP_036221.2 | 13 | 2586 | |
XM_011544135.2 | XP_011542437.1 | 13 | 3357 | ||
XM_011544136.2 | XP_011542438.1 | 11 | 1823 | ||
GenBank mRNA | AB013380.1 | - | 13 | 2598 | |
AF216833.1 | - | 13 | 2586 | ||
AK172794.1 | - | 12 | 1843 | ||
BC064930.1 | - | 13 | 2581 | 6143235 |