Gene Transcripts

Gene Symbol
SLC2A6
Entrez Gene ID
11182
Gene Name
solute carrier family 2 member 6
Gene Aliases
GLUT6, GLUT9, HSA011372
Chromosome Location
Chr. 9: 133471094 - 133479154 on Build GRCh38
UniGene ID
Hs.244378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145099.1 NP_001138571.1 3 435
NM_017585.3 NP_060055.2 3 435
XM_011518189.2 XP_011516491.2 3 408
XM_017014236.1 XP_016869725.1 3 417
XM_017014237.1 XP_016869726.1 3 408
XM_017014238.1 XP_016869727.1 3 408
GenBank mRNA AB209108.1 - 2 700
AJ011372.2 - 3 377
AK074836.1 - 3 385
AK074927.1 - 3 418
AK096630.1 - 2 938
AK222919.1 - 3 380
AK225106.1 - 3 377
AK294974.1 - 3 355
BC013740.1 - 3 369 4449594
DC305025.1 - 3 435
Y17803.1 - 3 386

View Full Product Details