Gene Transcripts

Gene Symbol
SLC17A7
Entrez Gene ID
57030
Gene Name
solute carrier family 17 member 7
Gene Aliases
BNPI, VGLUT1
Chromosome Location
Chr. 19: 49429398 - 49441551 on Build GRCh38
UniGene ID
Hs.375616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020309.3 NP_064705.1 12 1977
GenBank mRNA AB032436.1 - 12 1826
AK091061.1 - 8 1146
AK289622.1 - 9 1385
AK294226.1 - 12 1783
AK294405.1 - 12 1826
BC059379.1 - 12 1952 5262415

View Full Product Details