Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199148.1 | NP_001186077.1 | 8 | 873 | |
NM_173512.2 | NP_775783.1 | 7 | 807 | ||
XM_005246350.4 | XP_005246407.1 | 9 | 1026 | ||
XM_011510737.2 | XP_011509039.1 | 8 | 1118 | ||
XM_011510742.2 | XP_011509044.1 | 6 | 814 | ||
XM_017003455.1 | XP_016858944.1 | 9 | 1184 | ||
XM_017003456.1 | XP_016858945.1 | 8 | 1118 | ||
XM_017003457.1 | XP_016858946.1 | 8 | 1109 | ||
XM_017003458.1 | XP_016858947.1 | 8 | 1069 | ||
XM_017003459.1 | XP_016858948.1 | 7 | 807 | ||
XM_017003460.1 | XP_016858949.1 | 9 | 1184 | ||
XM_017003461.1 | XP_016858950.1 | 9 | 1184 | ||
XM_017003462.1 | XP_016858951.1 | 9 | 1184 | ||
XM_017003463.1 | XP_016858952.1 | 8 | 659 | ||
XM_017003464.1 | XP_016858953.1 | 5 | 793 | ||
XM_017003465.1 | XP_016858954.1 | 4 | 321 | ||
GenBank mRNA | AK097141.1 | - | 7 | 807 | |
AK293998.1 | - | 8 | 834 | ||
BC125157.1 | - | 10 | 800 | 40120878 |