Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018897.2 | NP_061720.2 | 9 | 900 | |
XM_011511487.2 | XP_011509789.1 | 10 | 1276 | ||
XM_011511488.2 | XP_011509790.1 | 9 | 2070 | ||
XM_011511489.2 | XP_011509791.1 | 8 | 862 | ||
XM_011511490.2 | XP_011509792.1 | 9 | 2070 | ||
XM_011511491.2 | XP_011509793.1 | 9 | 2072 | ||
XM_011511492.2 | XP_011509794.1 | 9 | 2072 | ||
XM_011511493.2 | XP_011509795.1 | 9 | 2072 | ||
XM_011511494.2 | XP_011509796.1 | 9 | 2071 | ||
XM_011511495.2 | XP_011509797.1 | 9 | 2071 | ||
XM_017004504.1 | XP_016859993.1 | 9 | 2069 | ||
GenBank mRNA | AB023161.2 | - | 9 | 918 | |
AF327442.1 | - | 9 | 895 | ||
AK094515.1 | - | 9 | 900 |