Gene Transcripts
- Gene Symbol
- GGCX
- Entrez Gene ID
- 2677
- Gene Name
- gamma-glutamyl carboxylase
- Gene Aliases
- VKCFD1
- Chromosome Location
- Chr. 2: 85544720 - 85561534 on Build GRCh38
- UniGene ID
- Hs.77719
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000821.6 |
NP_000812.2
|
10, 11 |
1553 |
|
NM_001142269.3 |
NP_001135741.1
|
9, 10 |
1382 |
|
XM_005264259.4 |
XP_005264316.1
|
10, 11 |
1512 |
|
XM_011532764.2 |
XP_011531066.1
|
9, 10 |
1348 |
|
XM_011532765.2 |
XP_011531067.1
|
8, 9 |
1187 |
|
XM_017003803.1 |
XP_016859292.1
|
9, 10 |
1375 |
|
GenBank mRNA |
AK130139.1 |
- |
1, 2 |
145 |
|
AK222827.1 |
- |
10, 11 |
1513 |
|
AK225663.1 |
- |
10, 11 |
1512 |
|
AK297397.1 |
- |
9, 10 |
1322 |
|
AK313126.1 |
- |
10, 11 |
1521 |
|
BC013979.2 |
- |
10, 11 |
1475 |
4129593 |
EU847509.1 |
- |
10, 11 |
1450 |
|
L17128.1 |
- |
10, 11 |
1602 |
|
M81592.1 |
- |
10, 11 |
1533 |
|
View Full Product Details