Gene Transcripts

Gene Symbol
ACY1
Entrez Gene ID
95
Gene Name
aminoacylase 1
Gene Aliases
ACY-1, ACY1D, HEL-S-5
Chromosome Location
Chr. 3: 51983284 - 51989202 on Build GRCh38
UniGene ID
Hs.334707
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000666.2 NP_000657.1 11 1078
NM_001198895.1 NP_001185824.1 11 1073
NM_001198896.1 NP_001185825.1 9 862
NM_001198898.1 NP_001185827.1 10 973
GenBank mRNA AK298274.1 - 11 827
BC000545.2 - 11 822 3162222
BC003023.1 - 11 815 2821665
BC014112.2 - 11 798 4543442
CD014023.1 - 9 703
CD014027.1 - 10 814
D14524.1 - 11 812
D16307.1 - 11 810
EU794586.1 - 11 798
KU177890.1 - 10 754
L07548.1 - 11 815

Gene Symbol
ABHD14A-ACY1
Entrez Gene ID
100526760
Gene Name
ABHD14A-ACY1 readthrough
Gene Aliases
Chromosome Location
Chr. 3: 51975026 - 51989202 on Build GRCh38
UniGene ID
Hs.334707
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316331.1 NP_001303260.1 13 1279

View Full Product Details