Gene Transcripts

Gene Symbol
MANBA
Entrez Gene ID
4126
Gene Name
mannosidase beta
Gene Aliases
MANB1
Chromosome Location
Chr. 4: 102631486 - 102760998 on Build GRCh38
UniGene ID
Hs.480415
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005908.3 NP_005899.3 2 309
XM_017008203.1 XP_016863692.1 3 295
XM_017008204.1 XP_016863693.1 2 264
GenBank mRNA AB209846.1 - 2 256
AK291598.1 - 2 297
AK300015.1 - 2 250
AK312827.1 - 2 264
AU132611.1 - 2 309
BC015743.1 - 2 299 4905535
EU009130.1 - 2 209

View Full Product Details