Gene Transcripts

Gene Symbol
ATP8B3
Entrez Gene ID
148229
Gene Name
ATPase phospholipid transporting 8B3
Gene Aliases
ATPIK
Chromosome Location
Chr. 19: 1782070 - 1812276 on Build GRCh38
UniGene ID
Hs.306212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178002.2 NP_001171473.1 7 745
NM_138813.3 NP_620168.1 7 884
NR_047593.2 7 772
XM_006722654.2 XP_006722717.1 7 884
XM_006722655.2 XP_006722718.1 7 963
XM_006722656.2 XP_006722719.1 7 772
XM_011527707.1 XP_011526009.1 7 884
XM_011527708.1 XP_011526010.1 7 884
XM_011527709.1 XP_011526011.1 7 884
XM_011527711.2 XP_011526013.1 7 884
GenBank mRNA AK125225.1 - 7 666
AK125969.1 - 7 772
AY302538.1 - 7 708
BC035162.2 - 7 879 5265541

View Full Product Details