Gene Transcripts

Gene Symbol
SLC16A11
Entrez Gene ID
162515
Gene Name
solute carrier family 16 member 11
Gene Aliases
MCT11
Chromosome Location
Chr. 17: 7041617 - 7044411 on Build GRCh38
UniGene ID
Hs.336564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153357.1 NP_699188.1 3 901
XM_005256488.3 XP_005256545.2 4 1134
XM_017024281.1 XP_016879770.1 4 1134
GenBank mRNA AK074674.1 - 3 901
BC093860.1 - 3 584 7939705
BC093886.1 - 3 584 7939731

View Full Product Details