Gene Transcripts
- Gene Symbol
- RASGRF1
- Entrez Gene ID
- 5923
- Gene Name
- Ras protein specific guanine nucleotide releasing factor 1
- Gene Aliases
- CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
- Chromosome Location
- Chr. 15: 78959947 - 79090873 on Build GRCh38
- UniGene ID
- Hs.459035
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145648.1 |
NP_001139120.1
|
15, 16 |
2833 |
|
NM_002891.4 |
NP_002882.3
|
16, 17 |
2881 |
|
NM_153815.2 |
NP_722522.1
|
2, 3 |
1327 |
|
XM_011521866.2 |
XP_011520168.1
|
16, 17 |
2781 |
|
XM_011521867.2 |
XP_011520169.1
|
16, 17 |
2781 |
|
XM_017022455.1 |
XP_016877944.1
|
16, 17 |
2772 |
|
XM_017022456.1 |
XP_016877945.1
|
15, 16 |
2742 |
|
XM_017022457.1 |
XP_016877946.1
|
15, 16 |
2742 |
|
XM_017022458.1 |
XP_016877947.1
|
15, 16 |
2733 |
|
XM_017022459.1 |
XP_016877948.1
|
15, 16 |
2742 |
|
GenBank mRNA |
AK226101.1 |
- |
14, 15 |
2117 |
|
AK290135.1 |
- |
16, 17 |
2747 |
|
AK309047.1 |
- |
14, 15 |
2264 |
|
BC040275.1 |
- |
15, 16 |
2724 |
5192914 |
L26584.1 |
- |
16, 17 |
2512 |
|
S62035.1 |
- |
2, 3 |
1336 |
|
View Full Product Details