Gene Transcripts

Gene Symbol
UBE2G2
Entrez Gene ID
7327
Gene Name
ubiquitin conjugating enzyme E2 G2
Gene Aliases
UBC7
Chromosome Location
Chr. 21: 44768580 - 44801836 on Build GRCh38
UniGene ID
Hs.529420
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202489.1 NP_001189418.1 4 1508
NM_003343.5 NP_003334.2 6 1590
NM_182688.2 NP_872630.1 7 1705
XM_017028469.1 XP_016883958.1 6 1549
GenBank mRNA AF032456.1 - 6 1557
AK122700.1 - 2 3007
AL834129.1 - 2 2324
BC001738.1 - 6 1553 3536853
BC008351.1 - 6 1550 3536171

View Full Product Details