Gene Transcripts

Gene Symbol
SUCLA2
Entrez Gene ID
8803
Gene Name
succinate-CoA ligase ADP-forming beta subunit
Gene Aliases
A-BETA, MTDPS5, SCS-betaA
Chromosome Location
Chr. 13: 47942656 - 48037972 on Build GRCh38
UniGene ID
Hs.743361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003850.2 NP_003841.1 9, 10 1293
XM_017020806.1 XP_016876295.1 9, 10 1513
XM_017020807.1 XP_016876296.1 11, 12 1528
XM_017020809.1 XP_016876298.1 10, 11 1377
GenBank mRNA AB035863.1 - 9, 10 1293
AF058953.1 - 8, 9 1125
AF087890.1 - 9, 10 1301
AK001458.1 - 9, 10 1191
AK001847.1 - 9, 10 1280
AK093506.1 - 3, 4 1607
AK225452.1 - 9, 10 1280
AK299437.1 - 9, 10 1232
AK302311.1 - 10, 11 1352
AK315513.1 - 9, 10 1255
AK316041.1 - 10, 11 1207
AK316271.1 - 10, 11 1608
AL080096.2 - 4, 5 482
BC027587.2 - 9, 10 1245 4829291
BX640602.1 - 9, 10 1264
KU178442.1 - 9, 10 1236
KU178443.1 - 6, 7 792

View Full Product Details