Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004163.4 | NP_004154.2 | 6 | 1095 | |
XM_005266733.1 | XP_005266790.1 | 7 | 1016 | ||
XM_017025913.1 | XP_016881402.1 | 9 | 1849 | ||
XM_017025914.1 | XP_016881403.1 | 8 | 1244 | ||
GenBank mRNA | AF329499.1 | - | 6 | 1095 | |
BC027474.1 | - | 6 | 963 | 4520182 | |
U57093.2 | - | 6 | 943 |