Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002802.2 | NP_002793.2 | 4 | 264 | |
XM_017021470.1 | XP_016876959.1 | 4 | 1783 | ||
XM_017021471.1 | XP_016876960.1 | 5 | 340 | ||
GenBank mRNA | AB451311.1 | - | 1 | 216 | |
AB451441.1 | - | 1 | 216 | ||
AK222668.1 | - | 4 | 232 | ||
AK299121.1 | - | 3 | 140 | ||
AK313325.1 | - | 4 | 253 | ||
BC000512.2 | - | 1 | 250 | 2822718 | |
BC016368.1 | - | 1 | 244 | 4133348 | |
BC018859.2 | - | 1 | 217 | ||
BC073818.1 | - | 1 | 222 | 5456334 | |
BT009826.1 | - | 1 | 216 | ||
CR457044.1 | - | 1 | 216 | ||
L02426.1 | - | 4 | 276 |