Gene Transcripts
- Gene Symbol
- MDM4
- Entrez Gene ID
- 4194
- Gene Name
- MDM4, p53 regulator
- Gene Aliases
- HDMX, MDMX, MRP1
- Chromosome Location
- Chr. 1: 204516379 - 204558120 on Build GRCh38
- UniGene ID
- Hs.497492
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204171.1 |
NP_001191100.1
|
3, 4 |
327 |
|
NM_001278516.1 |
NP_001265445.1
|
3, 4 |
327 |
|
NM_001278518.1 |
NP_001265447.1
|
3, 4 |
327 |
|
NM_002393.4 |
NP_002384.2
|
3, 4 |
327 |
|
XM_017001311.1 |
XP_016856800.1
|
3, 4 |
326 |
|
XM_017001312.1 |
XP_016856801.1
|
3, 4 |
326 |
|
XM_017001313.1 |
XP_016856802.1
|
3, 4 |
326 |
|
GenBank mRNA |
AF007111.1 |
- |
3, 4 |
277 |
|
AK223228.1 |
- |
3, 4 |
290 |
|
AK303626.1 |
- |
3, 4 |
300 |
|
AK304698.1 |
- |
3, 4 |
300 |
|
AK310607.1 |
- |
3, 4 |
303 |
|
AK310903.1 |
- |
3, 4 |
300 |
|
AY949028.1 |
- |
2, 3 |
161 |
|
BC067299.1 |
- |
3, 4 |
317 |
30390159 |
BC105106.1 |
- |
3, 4 |
220 |
8144109 |
BC143431.1 |
- |
3, 4 |
220 |
|
BC143432.1 |
- |
3, 4 |
220 |
|
BP247507.1 |
- |
3, 4 |
327 |
|
BX640923.1 |
- |
3, 4 |
301 |
|
CR749470.1 |
- |
3, 4 |
322 |
|
KC479043.1 |
- |
2, 3 |
161 |
|
KF435149.1 |
- |
2, 3 |
161 |
|
KF435150.1 |
- |
2, 3 |
161 |
|
View Full Product Details