Gene Transcripts

Gene Symbol
CSTA
Entrez Gene ID
1475
Gene Name
cystatin A
Gene Aliases
AREI, PSS4, STF1, STFA
Chromosome Location
Chr. 3: 122325164 - 122341969 on Build GRCh38
UniGene ID
Hs.518198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005213.3 NP_005204.1 3 424
GenBank mRNA AK291308.1 - 3 340
AV717631.1 - 3 334
BC010379.1 - 3 344 4063525
BG213814.1 - 2 231
X05978.1 - 3 340

View Full Product Details