Gene Transcripts

Gene Symbol
XPNPEP3
Entrez Gene ID
63929
Gene Name
X-prolyl aminopeptidase 3
Gene Aliases
APP3, ICP55, NPHPL1
Chromosome Location
Chr. 22: 40857081 - 40932819 on Build GRCh38
UniGene ID
Hs.529163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022098.3 NP_071381.1 2 225
GenBank mRNA AK301635.1 - 2 224
AK310502.1 - 2 483
AK313770.1 - 2 186
AL365514.1 - 2 208
AL834310.1 - 2 175
BC001208.1 - 2 160 3502251
BC001681.2 - 2 192 2823656
BC004989.1 - 2 192 2823656
BP346902.1 - 2 229
BX648018.1 - 2 220
CR456442.1 - 2 161
CR457396.1 - 2 124

View Full Product Details