Gene Transcripts
- Gene Symbol
- NRBP2
- Entrez Gene ID
- 340371
- Gene Name
- nuclear receptor binding protein 2
- Gene Aliases
- TRG16, pp9320
- Chromosome Location
- Chr. 8: 143829776 - 143840974 on Build GRCh38
- UniGene ID
- Hs.521926
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178564.3 |
NP_848659.2
|
14, 15 |
1404 |
|
XM_017013378.1 |
XP_016868867.1
|
14, 15 |
1404 |
|
XM_017013379.1 |
XP_016868868.1
|
14, 15 |
1404 |
|
XM_017013380.1 |
XP_016868869.1
|
14, 15 |
1404 |
|
XM_017013381.1 |
XP_016868870.1
|
14, 15 |
1404 |
|
XM_017013382.1 |
XP_016868871.1
|
14, 15 |
1404 |
|
XM_017013383.1 |
XP_016868872.1
|
14, 15 |
1404 |
|
GenBank mRNA |
AF318376.1 |
- |
4, 5 |
744 |
|
AK094017.1 |
- |
1, 2 |
113 |
|
AK308967.1 |
- |
1, 2 |
95 |
|
AL137662.1 |
- |
12, 13 |
1333 |
|
AL834530.1 |
- |
7, 8 |
643 |
|
AY277601.1 |
- |
4, 5 |
731 |
|
BC071605.1 |
- |
13, 14 |
1567 |
|
BC113873.1 |
- |
11, 12 |
1408 |
40053695 |
BC114481.1 |
- |
12, 13 |
1305 |
|
CR457350.1 |
- |
6, 7 |
535 |
|
View Full Product Details