Gene Transcripts

Gene Symbol
WNT9A
Entrez Gene ID
7483
Gene Name
Wnt family member 9A
Gene Aliases
WNT14
Chromosome Location
Chr. 1: 227918658 - 227947975 on Build GRCh38
UniGene ID
Hs.149504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011544271.2 XP_011542573.1 4 1808
GenBank mRNA AB060283.1 - 4 1248

View Full Product Details