Gene Transcripts

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
Chromosome Location
Chr. 9: 130902438 - 130939068 on Build GRCh38
UniGene ID
Hs.133205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145106.1 NP_001138578.1 3 382
NM_032843.4 NP_116232.3 2 520
GenBank mRNA AK027716.1 - 1 136
AK292489.1 - 2 289
AK300287.1 - 374
AY358311.1 - 2 277
BC032953.2 - 2 518 4811679
BC112892.1 - 2 270
DC404353.1 - 3 376

View Full Product Details